Assay Details
Gene Symbol: LOC5576045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662320.2 | XP_001662370.2 | 6 - 7 | 1210 | 72 | |
XM_021842617.1 | XP_021698309.1 | 5 - 6 | 1182 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
