Assay Details
Gene Symbol: LOC5576419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841278.1 | XP_021696970.1 | 2 - 3 | 1030 | 96 | |
XM_021841279.1 | XP_021696971.1 | 2 - 3 | 1133 | 96 | ||
XM_021841280.1 | XP_021696972.1 | 2 - 3 | 1130 | 96 | ||
XM_021841281.1 | XP_021696973.1 | 2 - 3 | 982 | 96 | ||
XM_021841282.1 | XP_021696974.1 | 2 - 3 | 979 | 96 | ||
XM_021841284.1 | XP_021696976.1 | 2 - 3 | 871 | 96 | ||
XM_021841285.1 | XP_021696977.1 | 2 - 3 | 887 | 96 | ||
XM_021841286.1 | XP_021696978.1 | 2 - 3 | 866 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021841278.1, XM_021841279.1, XM_021841280.1, XM_021841281.1, XM_021841282.1, XM_021841284.1, XM_021841285.1, XM_021841286.1 transcripts.
Set Membership: |
![]() ![]() |
