Assay Details
Gene Symbol: LOC5570012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854804.1 | XP_021710496.1 | 5 - 6 | 1712 | 101 | |
XM_021854805.1 | XP_021710497.1 | 5 - 6 | 1762 | 101 | ||
XM_021854806.1 | XP_021710498.1 | 3 - 4 | 1184 | 101 |

More Information
Set Membership: |
![]() ![]() |
