Assay Details
Gene Symbol: LOC5570012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854804.1 | XP_021710496.1 | 4 - 5 | 1512 | 79 | |
XM_021854805.1 | XP_021710497.1 | 4 - 5 | 1562 | 79 | ||
XM_021854806.1 | XP_021710498.1 | 2 - 3 | 984 | 79 |
