Assay Details
Gene Symbol: LOC5570205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658937.2 | XP_001658987.1 | 3 - 4 | 1365 | 63 | |
XM_021852388.1 | XP_021708080.1 | 3 - 4 | 1239 | 63 | ||
XM_021852389.1 | XP_021708081.1 | 3 - 4 | 1443 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
