Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 4 - 5 | 2755 | 58 | |
XM_021853023.1 | XP_021708715.1 | 2 - 3 | 2239 | 58 | ||
XM_021853024.1 | XP_021708716.1 | 2 - 3 | 2187 | 58 | ||
XM_021853027.1 | XP_021708719.1 | 4 - 5 | 2752 | 58 | ||
XM_021853028.1 | XP_021708720.1 | 4 - 5 | 2762 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021853022.1, XM_021853023.1, XM_021853024.1, XM_021853027.1, XM_021853028.1 transcripts.
Set Membership: |
![]() ![]() |
