Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 11 - 12 | 5114 | 69 | |
XM_021853023.1 | XP_021708715.1 | 9 - 10 | 4598 | 69 | ||
XM_021853024.1 | XP_021708716.1 | 9 - 10 | 4546 | 69 | ||
XM_021853025.1 | XP_021708717.1 | 6 - 7 | 2364 | 69 | ||
XM_021853027.1 | XP_021708719.1 | 11 - 12 | 5111 | 69 | ||
XM_021853028.1 | XP_021708720.1 | 11 - 12 | 5121 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
