Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 10 - 11 | 4987 | 72 | |
XM_021853023.1 | XP_021708715.1 | 8 - 9 | 4471 | 72 | ||
XM_021853024.1 | XP_021708716.1 | 8 - 9 | 4419 | 72 | ||
XM_021853025.1 | XP_021708717.1 | 5 - 6 | 2237 | 72 | ||
XM_021853027.1 | XP_021708719.1 | 10 - 11 | 4984 | 72 | ||
XM_021853028.1 | XP_021708720.1 | 10 - 11 | 4994 | 72 |
