Assay Details
Gene Symbol: LOC5572215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853022.1 | XP_021708714.1 | 9 - 10 | 4880 | 103 | |
XM_021853023.1 | XP_021708715.1 | 7 - 8 | 4364 | 103 | ||
XM_021853024.1 | XP_021708716.1 | 7 - 8 | 4312 | 103 | ||
XM_021853025.1 | XP_021708717.1 | 4 - 5 | 2130 | 103 | ||
XM_021853027.1 | XP_021708719.1 | 9 - 10 | 4877 | 103 | ||
XM_021853028.1 | XP_021708720.1 | 9 - 10 | 4887 | 103 |

More Information
Set Membership: |
![]() ![]() |
