Assay Details
Gene Symbol: LOC5575522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848029.1 | XP_021703721.1 | 2 - 3 | 600 | 67 | |
XM_021848030.1 | XP_021703722.1 | 2 - 3 | 587 | 67 | ||
XM_021848031.1 | XP_021703723.1 | 2 - 3 | 590 | 67 | ||
XM_021848032.1 | XP_021703724.1 | 2 - 3 | 575 | 67 | ||
XM_021848033.1 | XP_021703725.1 | 2 - 3 | 545 | 67 | ||
XM_021848034.1 | XP_021703726.1 | 3 - 4 | 621 | 67 | ||
XM_021848035.1 | XP_021703727.1 | 2 - 3 | 453 | 67 | ||
XM_021848036.1 | XP_021703728.1 | 3 - 4 | 529 | 67 | ||
XM_021848037.1 | XP_021703729.1 | 2 - 3 | 456 | 67 | ||
XM_021848039.1 | XP_021703731.1 | 1 - 2 | 273 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848029.1, XM_021848030.1, XM_021848031.1, XM_021848032.1, XM_021848033.1, XM_021848034.1, XM_021848035.1, XM_021848036.1, XM_021848037.1, XM_021848039.1 transcripts.
Set Membership: |
![]() ![]() |
