Assay Details
Gene Symbol: LOC5575522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848028.1 | XP_021703720.1 | 6 - 7 | 1333 | 81 | |
XM_021848029.1 | XP_021703721.1 | 5 - 6 | 1092 | 81 | ||
XM_021848030.1 | XP_021703722.1 | 5 - 6 | 1079 | 81 | ||
XM_021848031.1 | XP_021703723.1 | 5 - 6 | 1082 | 81 | ||
XM_021848032.1 | XP_021703724.1 | 5 - 6 | 1067 | 81 | ||
XM_021848033.1 | XP_021703725.1 | 5 - 6 | 1037 | 81 | ||
XM_021848034.1 | XP_021703726.1 | 6 - 7 | 1113 | 81 | ||
XM_021848035.1 | XP_021703727.1 | 5 - 6 | 945 | 81 | ||
XM_021848036.1 | XP_021703728.1 | 6 - 7 | 1021 | 81 | ||
XM_021848037.1 | XP_021703729.1 | 5 - 6 | 948 | 81 | ||
XM_021848039.1 | XP_021703731.1 | 4 - 5 | 765 | 81 | ||
XM_021848040.1 | XP_021703732.1 | 5 - 6 | 668 | 81 |
