Assay Details
Gene Symbol: LOC5575522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848028.1 | XP_021703720.1 | 7 - 8 | 1502 | 100 | |
XM_021848029.1 | XP_021703721.1 | 6 - 7 | 1261 | 100 | ||
XM_021848030.1 | XP_021703722.1 | 6 - 7 | 1248 | 100 | ||
XM_021848031.1 | XP_021703723.1 | 6 - 7 | 1251 | 100 | ||
XM_021848032.1 | XP_021703724.1 | 6 - 7 | 1236 | 100 | ||
XM_021848033.1 | XP_021703725.1 | 6 - 7 | 1206 | 100 | ||
XM_021848034.1 | XP_021703726.1 | 7 - 8 | 1282 | 100 | ||
XM_021848035.1 | XP_021703727.1 | 6 - 7 | 1114 | 100 | ||
XM_021848036.1 | XP_021703728.1 | 7 - 8 | 1190 | 100 | ||
XM_021848037.1 | XP_021703729.1 | 6 - 7 | 1117 | 100 | ||
XM_021848039.1 | XP_021703731.1 | 5 - 6 | 934 | 100 | ||
XM_021848040.1 | XP_021703732.1 | 6 - 7 | 837 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
