Assay Details
Gene Symbol: LOC5568734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842325.1 | XP_021698017.1 | 11 - 12 | 3310 | 101 | |
XM_021842326.1 | XP_021698018.1 | 11 - 12 | 3415 | 101 | ||
XM_021842327.1 | XP_021698019.1 | 11 - 12 | 3283 | 101 | ||
XM_021842329.1 | XP_021698021.1 | 11 - 12 | 3262 | 101 | ||
XM_021842330.1 | XP_021698022.1 | 11 - 12 | 3367 | 101 | ||
XM_021842331.1 | XP_021698023.1 | 10 - 11 | 2977 | 101 | ||
XM_021842332.1 | XP_021698024.1 | 10 - 11 | 3081 | 101 | ||
XM_021842333.1 | XP_021698025.1 | 10 - 11 | 3055 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
