Assay Details
Gene Symbol: LOC5568734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842325.1 | XP_021698017.1 | 5 - 6 | 1503 | 70 | |
XM_021842326.1 | XP_021698018.1 | 5 - 6 | 1620 | 70 | ||
XM_021842329.1 | XP_021698021.1 | 5 - 6 | 1503 | 70 | ||
XM_021842330.1 | XP_021698022.1 | 5 - 6 | 1620 | 70 | ||
XM_021842331.1 | XP_021698023.1 | 5 - 6 | 1503 | 70 | ||
XM_021842332.1 | XP_021698024.1 | 5 - 6 | 1619 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
