Assay Details
Gene Symbol: LOC5573478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654575.2 | XP_001654625.2 | 1 - 2 | 304 | 86 | |
XM_021840323.1 | XP_021696015.1 | 1 - 2 | 298 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
