Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 3 - 4 | 1371 | 64 | |
XM_021837840.1 | XP_021693532.1 | 3 - 4 | 1439 | 64 | ||
XM_021837841.1 | XP_021693533.1 | 3 - 4 | 1374 | 64 | ||
XM_021837842.1 | XP_021693534.1 | 4 - 5 | 1240 | 64 | ||
XM_021837843.1 | XP_021693535.1 | 3 - 4 | 1062 | 64 | ||
XM_021837844.1 | XP_021693536.1 | 3 - 4 | 1367 | 64 | ||
XM_021837845.1 | XP_021693537.1 | 3 - 4 | 1367 | 64 | ||
XM_021837846.1 | XP_021693538.1 | 3 - 4 | 1366 | 64 | ||
XM_021837847.1 | XP_021693539.1 | 3 - 4 | 1369 | 64 |

More Information
Set Membership: |
![]() ![]() |
