Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 1 - 2 | 529 | 96 | |
XM_021837840.1 | XP_021693532.1 | 1 - 2 | 529 | 96 | ||
XM_021837841.1 | XP_021693533.1 | 1 - 2 | 529 | 96 | ||
XM_021837842.1 | XP_021693534.1 | 2 - 3 | 398 | 96 | ||
XM_021837844.1 | XP_021693536.1 | 1 - 2 | 525 | 96 | ||
XM_021837845.1 | XP_021693537.1 | 1 - 2 | 525 | 96 | ||
XM_021837846.1 | XP_021693538.1 | 1 - 2 | 524 | 96 | ||
XM_021837847.1 | XP_021693539.1 | 1 - 2 | 524 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021837839.1, XM_021837840.1, XM_021837841.1, XM_021837842.1, XM_021837844.1, XM_021837845.1, XM_021837846.1, XM_021837847.1 transcripts.
Set Membership: |
![]() ![]() |
