Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 13 - 14 | 5426 | 110 | |
XM_021837840.1 | XP_021693532.1 | 13 - 14 | 5494 | 110 | ||
XM_021837841.1 | XP_021693533.1 | 13 - 14 | 5429 | 110 | ||
XM_021837842.1 | XP_021693534.1 | 14 - 15 | 5295 | 110 | ||
XM_021837843.1 | XP_021693535.1 | 13 - 14 | 5117 | 110 | ||
XM_021837844.1 | XP_021693536.1 | 14 - 15 | 3544 | 110 | ||
XM_021837845.1 | XP_021693537.1 | 13 - 14 | 3517 | 110 | ||
XM_021837846.1 | XP_021693538.1 | 12 - 13 | 3255 | 110 | ||
XM_021837847.1 | XP_021693539.1 | 12 - 13 | 3258 | 110 | ||
XM_021837848.1 | XP_021693540.1 | 6 - 7 | 1353 | 110 | ||
XM_021837850.1 | XP_021693542.1 | 5 - 6 | 1326 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
