Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 11 - 12 | 4989 | 80 | |
XM_021837840.1 | XP_021693532.1 | 11 - 12 | 5057 | 80 | ||
XM_021837841.1 | XP_021693533.1 | 11 - 12 | 4992 | 80 | ||
XM_021837842.1 | XP_021693534.1 | 12 - 13 | 4858 | 80 | ||
XM_021837843.1 | XP_021693535.1 | 11 - 12 | 4680 | 80 | ||
XM_021837844.1 | XP_021693536.1 | 12 - 13 | 3107 | 80 | ||
XM_021837845.1 | XP_021693537.1 | 11 - 12 | 3080 | 80 | ||
XM_021837846.1 | XP_021693538.1 | 10 - 11 | 2818 | 80 | ||
XM_021837847.1 | XP_021693539.1 | 10 - 11 | 2821 | 80 | ||
XM_021837848.1 | XP_021693540.1 | 4 - 5 | 916 | 80 | ||
XM_021837850.1 | XP_021693542.1 | 3 - 4 | 889 | 80 |
