Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 9 - 10 | 4616 | 103 | |
XM_021837840.1 | XP_021693532.1 | 9 - 10 | 4684 | 103 | ||
XM_021837841.1 | XP_021693533.1 | 9 - 10 | 4619 | 103 | ||
XM_021837842.1 | XP_021693534.1 | 10 - 11 | 4485 | 103 | ||
XM_021837843.1 | XP_021693535.1 | 9 - 10 | 4307 | 103 |

More Information
Set Membership: |
![]() ![]() |
