Assay Details
Gene Symbol: LOC5564565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837839.1 | XP_021693531.1 | 7 - 8 | 2270 | 80 | |
XM_021837840.1 | XP_021693532.1 | 7 - 8 | 2338 | 80 | ||
XM_021837841.1 | XP_021693533.1 | 7 - 8 | 2273 | 80 | ||
XM_021837842.1 | XP_021693534.1 | 8 - 9 | 2139 | 80 | ||
XM_021837843.1 | XP_021693535.1 | 7 - 8 | 1961 | 80 | ||
XM_021837844.1 | XP_021693536.1 | 7 - 8 | 2266 | 80 | ||
XM_021837845.1 | XP_021693537.1 | 7 - 8 | 2266 | 80 | ||
XM_021837846.1 | XP_021693538.1 | 7 - 8 | 2265 | 80 | ||
XM_021837847.1 | XP_021693539.1 | 7 - 8 | 2268 | 80 |
