Assay Details
Gene Symbol: LOC5578658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848819.1 | XP_021704511.1 | 5 - 6 | 1624 | 84 | |
XM_021848829.1 | XP_021704521.1 | 5 - 6 | 1624 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
