Assay Details
Gene Symbol: LOC5574699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855282.1 | XP_021710974.1 | 4 - 5 | 1979 | 111 | |
XM_021855283.1 | XP_021710975.1 | 4 - 5 | 950 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
