Assay Details
Gene Symbol: LOC5574699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855282.1 | XP_021710974.1 | 5 - 6 | 2162 | 77 | |
XM_021855283.1 | XP_021710975.1 | 5 - 6 | 1133 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
