Assay Details
Gene Symbol: LOC5574699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855282.1 | XP_021710974.1 | 9 - 10 | 3136 | 119 | |
XM_021855283.1 | XP_021710975.1 | 9 - 10 | 2107 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
