Assay Details
Gene Symbol: LOC5567583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651776.2 | XP_001651826.2 | 1 - 2 | 138 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001651776.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
