Assay Details
Gene Symbol: LOC5577537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656499.2 | XP_001656549.1 | 3 - 4 | 704 | 64 | |
XM_001656500.3 | XP_001656550.2 | 3 - 4 | 427 | 64 | ||
XM_021847113.1 | XP_021702805.1 | 3 - 4 | 450 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
