Assay Details
Gene Symbol: LOC5577537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656499.2 | XP_001656549.1 | 2 - 3 | 526 | 93 | |
XM_001656500.3 | XP_001656550.2 | 2 - 3 | 249 | 93 | ||
XM_021847113.1 | XP_021702805.1 | 2 - 3 | 272 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
