Assay Details
Gene Symbol: LOC5574617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857661.1 | XP_021713353.1 | 12 - 13 | 4573 | 86 | |
XM_021857662.1 | XP_021713354.1 | 12 - 13 | 4570 | 86 | ||
XM_021857663.1 | XP_021713355.1 | 11 - 12 | 4498 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
