Assay Details
Gene Symbol: LOC5580164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837239.1 | XP_021692931.1 | 6 - 7 | 1260 | 72 | |
XM_021837240.1 | XP_021692932.1 | 6 - 7 | 1414 | 72 | ||
XM_021837241.1 | XP_021692933.1 | 6 - 7 | 1221 | 72 |
