Assay Details
Gene Symbol: LOC5568975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652577.2 | XP_001652627.2 | 2 - 3 | 1560 | 65 | |
XM_021854032.1 | XP_021709724.1 | 3 - 4 | 1503 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
