Assay Details
Gene Symbol: LOC5579092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850433.1 | XP_021706125.1 | 5 - 6 | 1563 | 74 | |
XM_021850434.1 | XP_021706126.1 | 4 - 5 | 1662 | 74 | ||
XM_021850435.1 | XP_021706127.1 | 4 - 5 | 1663 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
