Assay Details
Gene Symbol: LOC5568034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844513.1 | XP_021700205.1 | 10 - 11 | 1440 | 80 | |
XM_021844514.1 | XP_021700206.1 | 10 - 11 | 1594 | 80 | ||
XM_021844515.1 | XP_021700207.1 | 10 - 11 | 1541 | 80 | ||
XM_021844517.1 | XP_021700209.1 | 10 - 11 | 1413 | 80 | ||
XM_021844518.1 | XP_021700210.1 | 10 - 11 | 1336 | 80 | ||
XM_021844519.1 | XP_021700211.1 | 10 - 11 | 1336 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
