Assay Details
Gene Symbol: LOC5568034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844513.1 | XP_021700205.1 | 7 - 8 | 1158 | 86 | |
XM_021844514.1 | XP_021700206.1 | 7 - 8 | 1312 | 86 | ||
XM_021844515.1 | XP_021700207.1 | 7 - 8 | 1259 | 86 | ||
XM_021844517.1 | XP_021700209.1 | 7 - 8 | 1131 | 86 | ||
XM_021844518.1 | XP_021700210.1 | 7 - 8 | 1054 | 86 | ||
XM_021844519.1 | XP_021700211.1 | 7 - 8 | 1054 | 86 |
