Assay Details
Gene Symbol: LOC5568034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844513.1 | XP_021700205.1 | 2 - 3 | 408 | 103 | |
XM_021844514.1 | XP_021700206.1 | 2 - 3 | 562 | 103 | ||
XM_021844515.1 | XP_021700207.1 | 2 - 3 | 509 | 103 | ||
XM_021844517.1 | XP_021700209.1 | 2 - 3 | 381 | 103 | ||
XM_021844518.1 | XP_021700210.1 | 2 - 3 | 304 | 103 | ||
XM_021844519.1 | XP_021700211.1 | 2 - 3 | 304 | 103 |

More Information
Set Membership: |
![]() ![]() |
