Assay Details
Gene Symbol: LOC5573468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001660839.2 | XP_001660889.2 | 2 - 3 | 1696 | 73 | |
XM_021837361.1 | XP_021693053.1 | 2 - 3 | 1697 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
