Assay Details
Gene Symbol: LOC5574270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843260.1 | XP_021698952.1 | 6 - 7 | 3775 | 63 | |
XM_021843262.1 | XP_021698954.1 | 6 - 7 | 3810 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
