Assay Details
Gene Symbol: LOC5564266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851107.1 | XP_021706799.1 | 4 - 5 | 1925 | 86 | |
XM_021851108.1 | XP_021706800.1 | 4 - 5 | 1925 | 86 |
