Assay Details
Gene Symbol: LOC5564266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851107.1 | XP_021706799.1 | 5 - 6 | 2026 | 68 | |
XM_021851108.1 | XP_021706800.1 | 5 - 6 | 2026 | 68 |

More Information
Set Membership: |
![]() ![]() |
