Assay Details
Gene Symbol: LOC5564266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851107.1 | XP_021706799.1 | 6 - 7 | 2350 | 100 | |
XM_021851108.1 | XP_021706800.1 | 6 - 7 | 2350 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
