Assay Details
Gene Symbol: LOC5579330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854496.1 | XP_021710188.1 | 1 - 2 | 198 | 109 | |
XM_021854498.1 | XP_021710190.1 | 1 - 2 | 198 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021854496.1, XM_021854498.1 transcripts.
Set Membership: |
![]() ![]() |
