Assay Details
Gene Symbol: LOC5579330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854496.1 | XP_021710188.1 | 2 - 3 | 2198 | 79 | |
XM_021854497.1 | XP_021710189.1 | 2 - 3 | 2215 | 79 | ||
XM_021854498.1 | XP_021710190.1 | 2 - 3 | 2198 | 79 |
