Assay Details
Gene Symbol: LOC5579330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854496.1 | XP_021710188.1 | 3 - 4 | 2382 | 74 | |
XM_021854497.1 | XP_021710189.1 | 3 - 4 | 2399 | 74 | ||
XM_021854498.1 | XP_021710190.1 | 3 - 4 | 2382 | 74 |
