Assay Details
Gene Symbol: LOC5579330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854496.1 | XP_021710188.1 | 4 - 5 | 2959 | 71 | |
XM_021854497.1 | XP_021710189.1 | 4 - 5 | 2976 | 71 | ||
XM_021854498.1 | XP_021710190.1 | 4 - 5 | 2959 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
