Assay Details
Gene Symbol: LOC5571700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659794.2 | XP_001659844.2 | 4 - 5 | 2804 | 59 | |
XM_021842003.1 | XP_021697695.1 | 4 - 5 | 2788 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
