Assay Details
Gene Symbol: LOC5572351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841121.1 | XP_021696813.1 | 6 - 7 | 2328 | 74 | |
XM_021841122.1 | XP_021696814.1 | 7 - 8 | 1958 | 74 | ||
XM_021841123.1 | XP_021696815.1 | 7 - 8 | 1849 | 74 | ||
XM_021841125.1 | XP_021696817.1 | 6 - 7 | 2327 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
