Assay Details
Gene Symbol: LOC5572351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841121.1 | XP_021696813.1 | 1 - 2 | 1672 | 75 | |
XM_021841122.1 | XP_021696814.1 | 2 - 3 | 1302 | 75 | ||
XM_021841123.1 | XP_021696815.1 | 2 - 3 | 1193 | 75 | ||
XM_021841125.1 | XP_021696817.1 | 1 - 2 | 1671 | 75 |
