Assay Details
Gene Symbol: LOC5577228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854044.1 | XP_021709736.1 | 8 - 9 | 2476 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5577228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854044.1 | XP_021709736.1 | 8 - 9 | 2476 | 58 |
Set Membership: |
![]() ![]() ![]() ![]() |