Assay Details
Gene Symbol: LOC5574681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845958.1 | XP_021701650.1 | 3 - 4 | 436 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5574681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845958.1 | XP_021701650.1 | 3 - 4 | 436 | 73 |
Set Membership: |
![]() ![]() ![]() |