Assay Details
Gene Symbol: LOC5576284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857493.1 | XP_021713185.1 | 4 - 5 | 2406 | 86 | |
XM_021857494.1 | XP_021713186.1 | 4 - 5 | 2406 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
